missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris SDS PAGE Running Buffer, FASTRun, 10X
Compatible with Tris-Glycine SDS (SDS-PAGE) chemistry whether you buy premade or pour your own (PYO) polyacrylamide gels
87.89 € - 112.00 €
Specifications
CAS | 151-21-3 |
---|---|
Color | Colorless to Yellow |
pH Range | 7.7 to 8.3 (1X/10X soln. at 25°C) |
Form | Liquid |
Description
- Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
- Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.
Specifications
151-21-3 | |
Colorless to Yellow | |
7.7 to 8.3 (1X/10X soln. at 25°C) | |
Liquid |