Learn More
Thermo Scientific Chemicals TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8.3
86.33 € - 159.80 €
Specifications
Chemical Name or Material | TRIS-Tricine-SDS running buffer (10X) |
---|---|
Physical Form | Liquid |
Packaging | Plastic bottle |
Solubility Information | It is soluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15400167
|
Thermo Scientific Chemicals
J60992.AP |
500 mL |
86.33 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410167
|
Thermo Scientific Chemicals
J60992.K2 |
1 L |
159.80 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris Tricine Gel Electrophoresis is routinely used for the separation of small proteins and peptides with a molecular weight of I to 100 kDa. It is used for protein analysis. This type of gel is perfectly suited for the separation and identification of proteins in samples containing only a few different proteins to samples containing complex mixtures (e.g. from cell extracts).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
TRIS-Tricine-SDS running buffer (10X) | |
Liquid | |
Plastic bottle | |
It is soluble in water. |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8.3
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]glycine, 2-amino-2-(hydroxymethyl)-1,3-propanediol, Sodium
RUO – Research Use Only