missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) phosphite, 92%, Thermo Scientific Chemicals

Product Code. 10341720
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795-31-9
Molecular Formula C9H27O3PSi3
Molecular Weight (g/mol) 298.54
MDL Number MFCD00015588
InChI Key VMZOBROUFBEGAR-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
PubChem CID 137213
IUPAC Name tris(trimethylsilyl) phosphite
SMILES C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Density 0.8930g/mL
Boiling Point 78.0°C to 81.0°C (8.3 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula P[OSi(CH3)3]3
Refractive Index 1.4080 to 1.4110 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.893
Solubility Information Solubility in water: reacts.
Formula Weight 298.54
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) phosphite
Show More Show Less
compliance-icons
Product Identifier
  • Tris(trimethylsilyl) phosphite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'