missing translation for 'onlineSavingsMsg'
Learn More
Tris(trimethylsilyl) phosphite, 95%, Thermo Scientific Chemicals Product Code.: 11363327

Tris(trimethylsilyl) phosphite, 95%, Thermo Scientific Chemicals

Product Code. 11363327
2 g
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy

Product Code. 11363327

Brand: Thermo Scientific Alfa Aesar 017159.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris(trimethylsilyl) phosphite is a reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1795-31-9
Molecular Formula C9H27O3PSi3
Molecular Weight (g/mol) 298.541
MDL Number MFCD00015588
InChI Key VMZOBROUFBEGAR-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
PubChem CID 137213
IUPAC Name tris(trimethylsilyl) phosphite
SMILES C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Density 0.893 g/mL
Boiling Point 80°C (8 mmHg)
Refractive Index 1.409
Quantity 2 g
Sensitivity Air and moisture sensitive
Solubility Information Sparingly soluble in water.
Formula Weight 298.54
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) phosphite

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU27K036 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number017159.02,017159.04
Lot NumberY14I042 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog Number017159.02,017159.04
Lot NumberP16I048 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog Number017159.02,017159.04
Lot NumberR03I046 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog Number017159.02,017159.04
Lot NumberW10H028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number017159.02,017159.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(trimethylsilyl) phosphite, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.