Learn More
Tris(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals
34.80 € - 435.00 €
Chemical Identifiers
CAS | 1873-77-4 |
---|---|
Molecular Formula | C9H28Si4 |
Molecular Weight (g/mol) | 248.66 |
MDL Number | MFCD00077893 |
InChI Key | SQMFULTZZQBFBM-UHFFFAOYSA-N |
Synonym | tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl |
IUPAC Name | 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane |
SMILES | C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400627
|
Thermo Scientific Alfa Aesar
B22457.03 |
1 g |
34.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410627
|
Thermo Scientific Alfa Aesar
B22457.06 |
5 g |
115.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420627
|
Thermo Scientific Alfa Aesar
B22457.14 |
25 g |
435.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.
Solubility
Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
1873-77-4 | |
248.66 | |
SQMFULTZZQBFBM-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane |
C9H28Si4 | |
MFCD00077893 | |
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl | |
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C |
Specifications
82°C to 84°C (12mmHg) | |
C9H28Si4 | |
UN1993 | |
55°C (131°F) | |
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl | |
SQMFULTZZQBFBM-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane | |
248.67 | |
1.49 | |
0.806 |
1873-77-4 | |
MFCD00077893 | |
1923953 | |
14,9757 | |
Miscible with pentane,ether,toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water. | |
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C | |
248.66 | |
97% | |
Air Sensitive | |
Tris(trimethylsilyl)silane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only