Learn More
Macherey-Nagel™ Tritest™ pH Paper Roll
Featuring three indicator zones for the highest accuracy reading. Macherey Nagel™ Tritest™ pH Paper Rolls give the user the most precise readings possible in pH measurement.
Brand:  Macherey-Nagel™ 90501
Product Code. 10433151
  • 16.05 € / 1 roll
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Provides rapid and reliable results
  • Shows three different colors for each full pH unit
  • Specially equipped with a hydrophobic barrier between different indicators
  • Length: 5 m
Specifications
Specifications
1 to 11
For most precise reading these papers show three different colors for each full pH unit
1 roll, 5m length
pH Paper
10 mm
Roll
1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11
1 Each
5 m