Learn More
Thermo Scientific Chemicals Triton∣r X-100 lysis buffer, pH 7.4
70.25 € - 109.55 €
Specifications
Content And Storage | 2° to 8°C |
---|---|
Format | Liquid |
Product Type | Cell Lysis Reagent |
pH | 7.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424699
|
Thermo Scientific Chemicals
J62289.AK |
250mL |
70.25 €
250mL |
In Stock
Log in to see stock. |
|||||
15434699
|
Thermo Scientific Chemicals
J62289.AP |
500mL |
109.55 €
500mL |
In Stock
Log in to see stock. |
|||||
Description
It is widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions. Triton X-100 is a commonly used detergent in laboratories. Triton X-100 is widely used to lyse cells to extract protein or organelles, or to permeabilize the membranes of living cells. Apart from laboratory use, Triton X-100 can be found in several types of cleaning compounds,[8] ranging from heavy-duty industrial products to gentle detergents. It is also a popular ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol. It is a good micellar catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2° to 8°C | |
Liquid | |
Cell Lysis Reagent | |
7.4 |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only