missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triton™ X-100 Surfact-Amps™ Detergent Solution

Thermo Scientific™ Surfact-Amps X-100 Detergent Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

177.00 € - 1916.00 €

Specifications

Form Liquid
Product Line Surfact-Amps™, Triton™
Reagent Type Detergent Solution
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13444259
View Documents
Thermo Scientific™
85111
50 mL
177.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
11891445
View Documents
Thermo Scientific™
28314
6 x 10 mL
187.00 €
Pack of 6
In Stock
to see stock.
Add to basket
 
13454259
View Documents
Thermo Scientific™
85112
250 mL
532.00 €
250mL
In Stock
to see stock.
Add to basket
 
11358311
View Documents
Thermo Scientific™
28313
1 L
1916.00 €
1000mL
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Triton X-100 Detergent Surfact-Amps Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Triton™ X-100 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Triton X-100 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

  • Triton X-100 – a nonionic detergent for use in various protein methods

  • Accurate – precise 10% detergent solution in ultrapure water

  • Easy-to-use – solution is simple to dispense and dilute for use

  • Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

  • Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 647
  • Detergent Class: Nonionic
  • Aggregation Number: 140
  • Micelle Molecular Weight: 90,000
  • Critical Micelle Concentration (CMC): 0.24mM (0.0155%, w/v)
  • Cloud Point: 64°C
  • Dialyzable: No

Specifications

Specifications

Liquid
Surfact-Amps™, Triton™
Detergent Solution
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Triton X-100 Surfact-Amps Detergent Solution, 50 mL.

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Octylphenol ethoxylate

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.