Learn More
Trolox(R), 97%, Thermo Scientific Chemicals
55.70 € - 608.00 €
Chemical Identifiers
CAS | 53188-07-1 |
---|---|
Molecular Formula | C14H18O4 |
Molecular Weight (g/mol) | 250.29 |
MDL Number | MFCD00006846 |
InChI Key | GLEVLJDDWXEYCO-UHFFFAOYNA-N |
Synonym | trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm |
PubChem CID | 40634 |
ChEBI | CHEBI:82625 |
IUPAC Name | 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid |
SMILES | CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10454155
|
Thermo Scientific Acros
218940010 |
1 g | Glass bottle |
55.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782831
|
Thermo Scientific Acros
218940050 |
5 g | Glass bottle |
182.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10002910
|
Thermo Scientific Acros
218940250 |
25 g | Glass bottle |
608.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
53188-07-1 | |
250.29 | |
GLEVLJDDWXEYCO-UHFFFAOYNA-N | |
40634 | |
6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid |
C14H18O4 | |
MFCD00006846 | |
trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm | |
CHEBI:82625 | |
CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O |
Specifications
53188-07-1 | |
Beige to White | |
97% | |
MFCD00006846 | |
trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm | |
GLEVLJDDWXEYCO-UHFFFAOYNA-N | |
6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid | |
40634 | |
250.29 | |
Crystalline Powder |
187.0°C to 194.0°C | |
Authentic | |
C14H18O4 | |
18, V,7, 335 | |
Solubility in water: slightly soluble. Other solubilities: soluble in methanol, ethanol | |
CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O | |
250.29 | |
CHEBI:82625 | |
97% | |
Trolox*, 97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 258-422-8
missing translation for 'rtecsNumber' : DJ2273000
missing translation for 'tsca' : TSCA
- Trolox
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only