missing translation for 'onlineSavingsMsg'
Learn More

Tropolone, 98%, Thermo Scientific Chemicals

Product Code. 10533881
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004158
InChI Key MDYOLVRUBBJPFM-UHFFFAOYSA-N
Synonym tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
PubChem CID 10789
ChEBI CHEBI:79966
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES C1=CC=C(C(=O)C=C1)O

Specifications

Melting Point 50.0°C to 54.0°C
Color Beige-Cream to White
Boiling Point 115.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 9963
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethyl ether, acetone, organic solvents
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tropolone
Show More Show Less
Product Identifier
  • Tropolone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.