Learn More
Invitrogen™ Troponin T (TNNT1) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHTNNT1X5
Description
The Human Troponin T (TNNT1) ELISA quantitates Hu TNNT1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TNNT1. Principle of the method The Human TNNT1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Mutations in this gene have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for this gene undergo alternative splicing that results in many tissue-specific isoforms, however, the full-length nature of some of these variants has not yet been determined.Specifications
P13805 | |
0.35 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
7138 | |
<12% | |
Wash Buffer, Adhesive Plate Covers, Assay Diluent Concentrate, Pre-coated 96 well plates, Chromogen, Standard, Biotinylated Detection Antibody, Stop Solution, SAV-HRP | |
5 x 96 Tests | |
Cardiovascular Research | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.391 ng/mL to 25 ng/mL | |
0.35 ng/mL to 25 ng/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-9, Angiopoietin 2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN-gamma, IGF-2, IGF-2 R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Neprilysin, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, WIF-1 | |
Human | |
Colorimetric Microplate Reader | |
ANM,NEM5,STNT,TNT,TNTS | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
ANM, NEM5, STNT, TNT, TNTS | |
1 hr. 20 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.