missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Trypanosoma Brucei Antigen Microarray
Trypanosoma Brucei Antigen Microarray with 1522 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.022.001
This item is not returnable.
View return policy
Description
- Microarray Content: Three selected antigens of Trypanosoma brucei: LiTat 1.3, LiTat 1.5, MiTat 1.4
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 1522 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
2 array copies per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |