missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Trypsin (0.25%), phenol red
Trypsin (0.25%), phenol red
23.36 € - 425.00 €
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | –5 to –20°C |
Cell Type | Mammalian Cells |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520626
|
Gibco™
25050014 |
100 mL |
23.36 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11530626
|
Gibco™
25050030 |
20 x 100mL |
425.00 €
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:Quality Testing
Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance. In addition, prior to manufacturing, the raw materials are tested for endotoxin, Mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis.
Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.
cGMP Manufacturing and Quality System
Gibco™ Trypsin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Trypsin solution made in our Grand Island, New York facility (15050-065). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
Mammalian Cells | |
Liquid | |
Phenol Red | |
Mammalian cell culture | |
Porcine pancreas |
–5 to –20°C | |
1 X | |
Cell Culture Dissociation Reagent | |
No EDTA | |
For research use only |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trypsin (1X), liquid
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : trypsin
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title