missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Trypsin (1:250), powder
Trypsin (1:250), powder
Brand: Gibco™ 27250018
This item is not returnable.
View return policy
Description
Gibco™ Trypsin 1:250 powder is an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging, and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:Quality Testing
Gibco™ Trypsin 1:250 is tested for endotoxin, mycoplasma, bacterial, fungal and viral contamination. Other quality tests include multiple activity assays, ash analysis and moisture analysis.
Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.
cGMP Manufacturing and Quality System
For supply chain continuity, we repack Gibco™ trypsin at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
Specifications
No EDTA | |
Mammalian Cells | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
Low | |
Gibco™ | |
Room Temperature | |
In vitro Bioassay |
Storage conditions: 2–8°C. Shipping conditions: Ambient. Shelf life: 24 months from date of manufacture. |
|
250 X | |
Powder | |
No Phenol Red | |
Securitainer Capped Vial | |
100 g | |
Porcine Pancreas |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trypsin (1:250), powder
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Trypsin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction