Learn More
Thermo Scientific™ Tryptic Digestion Kits
Shop All Thermo Scientific ProductsDescription
Accurate identification and analysis of proteins is a central component of proteomic strategies for studying cellular functions and processes. Using mass spectrometry, proteins and relevant post-translational modifications can be accurately identified, but the method is highly dependent on the quality of the sample preparation. Thermo Scientific Pierce Digestion Kits provide enzymes and reagents for efficient, reproducible digestion and guanidination to improve peptide fragment ionization.
Pierce In-Solution Tryptic Digestion and Guanidination Kit
The Pierce In-Solution Tryptic Digestion and Guanidination Kit provides an optimized procedure and reagents for approximately 90 digests, each containing 0.025 to 10 μg of protein.
• Complete digestion—trypsin completely digests protein ranging from 0.025 to 10 μg (∼90 reactions)
• Specific—guanidination reaction produces little to no N-terminal modification
• Convenient—kit includes reagents for digestion, reduction, alkylation, and guanidination
• Efficient—protein can be digested and guanidinated in a single day
Trypsin specifically cleaves peptide bonds at the carboxyl side of arginine and lysine residues. The signals from the arginine-containing peptides are generally stronger, due to the more basic side chain. To enhance overall ionization, guanidination converts lysines to homoarginines and is specific for the amine of lysine, but may occur minimally at the amine of the peptide's N-terminus. This derivatization leads to an increase in the intensity of the lysine-containing peptides and improved sequence coverage overall.
Pierce In-Gel Tryptic Digestion Kit
The Pierce In-Gel Tryptic Digestion Kit provides a complete set of reagents for performing approximately 150 digestions on colloidal Coomassie- or fluorescent dye-stained protein bands. The kit includes modified porcine trypsin, de-staining buffers, reduction reagents, alkylation reagents, and digestion buffers, along with detailed and simple instructions.
• Complete—includes all necessary reagents for gel destaining, reduction, alkylation, and tryptic digestion of proteins
• Robust—optimized procedure and reagents produce reliable digestions and data generation using a wide range of conditions
• Specific—contains highly purified and modified MS-grade trypsin with no chymotryptic activity and minimal autolytic activity
Two key sample preparation steps are the complete digestion of proteins into peptides and subsequent concentration and clean-up of peptides for mass spectrometry analysis. The Pierce In-Gel Tryptic Digestion Kit accomplishes the first of these steps for protein samples extracted from gel slices. Pierce C-18 Spin Columns and Tips accomplish the second step to clean-up the sample to be MS-ready.

Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Digestion and Guanidination Kit |
Content And Storage | Upon receipt, store the Pierce Trypsin Protease at -20°C in a nonfrost-free freezer; store other components at 4°C. |
Shipping Condition | Kit is shipped on gel pack. |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 90 Reactions |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
SPECS | Certificate of Analysis | 11/01/2023 | 89895 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.