missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soy Broth (Soybean Casein Digest Medium EP/USP/BP/JP) (Dehydrated)

Grow a wide variety of microorganisms with this general purpose medium containing two peptones.

Brand:  Thermo Scientific™ CM0129R

610.91 EUR valid until 2025-12-31
Use promo code "24336" to get your promotional price.


 View more versions of this product

Product Code. 11409658

  • 764.00 € / 2.50kg
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth EP/USP/BP/JP (Dehydrated). The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Use Tryptone Soya Broth to support the luxuriant growth of many fastidious organisms, without the addition of serum, while conforming to the formulations detailed in various international pharmacopoeias.

  • Suitable for tube dilution method of antibiotic susceptibility testing
  • Recommended in the USP for the recovery of organisms after sterilization processes4

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Dehydrated Culture Media
For 83.5L medium
Powder
2.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only