Learn More
Thermo Scientific™ Tryptone Soya Agar (Casein Soya Bean Digest Agar)
Grow a wide variety of microorganisms with general purpose Thermo Scientific™ Tryptone Soya Agar prepared media plates.
20.80 €
Specifications
Media Type Tryptone Soya Agar
Product Type Agar
Quantity 10/Pk.
View More Specs
Products 1
Product Code Brand Media Type Fill Volume Shelf Life Price Quantity & Availability  
Product Code Brand Media Type Fill Volume Shelf Life Price Quantity & Availability  
12932588
View Documents
Thermo Scientific™
PO0163A
Tryptone Soya Agar 19 mL 9 Weeks
20.80 €
Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
Grow a wide variety of microorganisms with Thermo Scientific ™ Tryptone Soya Agar. A general purpose agar medium, containing two peptones, Tryptone Soya Agar will support the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition.
Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopoeia and food testing methods for the growth of a wide variety of organisms. The product is tested to the pharmacopoeia and is mainly used as reference medium when testing selective media.
  • Has the ready-to-use convenience of prepared media
  • May be used as a blood agar base
  • Recommended as a reference medium for testing selective media
  • Conforms to formulations detailed in International Pharmacopoeia
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
Tryptone Soya Agar
Agar
10/Pk.
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 26 Weeks