Learn More
Thermo Scientific™ Tryptone Soya Broth (Bottled)
Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.
71.10 € - 75.90 €
Specifications
Color | Straw, Clear |
---|---|
Content And Storage | 2 to 25°C |
Manufacturing Origin | United Kingdom |
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fill Weight | Price | Quantity & Availability | |||||
12786959
|
Thermo Scientific™
BO0369E |
10.0g + 0.3g |
75.90 €
Pack of 24 |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
12693428
|
Thermo Scientific™
BO0369M |
100.0g + 1.5g |
71.10 €
Pack of 10 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.
- Conforms to formulations detailed in International Pharmacopoeia.
- Convenient: pre-filled bottles are ready for immediate use.
- Simple: easy to use with minimum training.
- Long shelf life: up to 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Straw, Clear | |
2 to 25°C | |
United Kingdom |
Safety and Handling
missing translation for 'shelfLife' : 52 Weeks