missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Freezers - Crated
GREENER_CHOICE

Product Code. 17117738 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
700 L
1500 L
Plug Type:
UK
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17117738

Brand: Thermo Scientific™ TSG1500FESCW

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from solid door 400 L to 1500 L double door.
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY

Specifications

Description TSG Series General Purpose Laboratory Freezer
Capacity (Metric) 1500 L
Temperature Range -12°C to -30°C
Temperature (Setpoint) -20°C
Door Style Solid
No. of Doors 2
Cabinet Material Steel
Interior Painted Steel
Shelves 8 Adjustable
Capacity (Metric) Shelf 30 kg
Certifications/Compliance CE
Casters Yes
Control Type Digital Microprocessor
Ports 1 Rear Access Port
Warranty 12 Months
Defrost Adaptive Defrost
Packaging Type Crated
Dimensions (D x W x H) Interior 690 x 1300 x 1510 mm
Dimensions (L x W x H) Exterior 690 x 1300 x 1510 mm
Monitoring Options Alarm
Smart View Compatibility Smart-Vue Pro Compatible
Plug Type UK
Voltage 230 V
Frequency 50 Hz
Green Features Energy Efficient
Refrigerant R290
Shipping Weight (Metric) 263 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.