Learn More
Thermo Scientific™ TSG General Purpose Refrigerators - Boxed
GREENER_CHOICE
The TSG Series of general purpose refrigerators are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Brand:  Thermo Scientific™ TSG1500REGBW
 View more versions of this product
Product Code. 17137738
  • 7305.00 € / Each
Explore more special offers
Description
Description
The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
  • Wide range of sizes from 400 L to 1500 L double door
  • Glass door (triple pane) and solid door configurations
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents Product Certifications
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TSG General Purpose Refrigerators - Boxed > TSG General Purpose Refrigerators, 1500 L, UK, Glass, Boxed