Learn More
TSPAN10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14430-1-AP-20UL
 View more versions of this product
Product Code. 16883064
  • 145.00 € / 20µL
Explore more special offers
Description
Description
TSPAN10 belongs to the tetraspanin (TM4SF) family. It is a multi-pass membrane protein. The exact function of TSPAN10 remains unknown.
Specifications
Specifications
TSPAN10
Polyclonal
Unconjugated
TSPAN10
OCSP, Oculospanin, tetraspanin 10, Tspan 10, TSPAN10
Rabbit
Antigen Affinity Chromatography
RUO
83882
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H1Z9
TSPAN10
TSPAN10 Fusion Protein Ag5807
20 μL