Learn More
TST™ Bowie Dick™ Test Pack
Test the steam penetration and air removal capability of a vacuum sterilizer with this test pack. The TST™ Bowie Dick Test Pack is a unique combination of advance Browne intelligent ink technology and the original Bowie Dick concept.
269.00 € - 450.00 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 11140-4(3) Class 2
For Use With (Application) For large porous load sterilizers
Product Type TST Bowie Dick Test Pack
Sterilization Indicator Yes
View More Specs
Products 2
Product Code Brand Sterilization Method Price Quantity & Availability  
Product Code Brand Sterilization Method Price Quantity & Availability  
12390489
View Documents
Steris Corporation
2310AB
Steam, Vacuum
269.00 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
12380489
View Documents
Steris Corporation
2352AB
Steam
450.00 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Single-use test pack
  • A successful test confirms that steam penetration into a test pack is rapid and even and that air and other non-condensable gases have been effectively removed
  • The chemical indicator sheet at the center of the pack shows a defined color change from yellow to dark blue/purple when exposed to a specific combination of time, temperature and steam
  • Conforms to ISO 11140-4 Class 2
Specifications
Specifications
Autoclavable
For large porous load sterilizers
Yes
ISO 11140-4(3) Class 2
TST Bowie Dick Test Pack
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
TST™ Bowie Dick™ Test Pack