Learn More
TST™ TST™ Control™ Self Adhesive Emulating Indicator Strip
Place the TST™ Control™ Self Adhesive Emulating Indicator Strip inside trays, packs or pouches when autoclaving so the indicator will confirm that good quality steam has penetrated to the point of placement.
Brand:  TST™ 2340AB
 View more versions of this product
Product Code. 12916996
  • 55.25 € / Pack of 100
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • ISO 11140 part 1, class 6 classification
  • Paper
  • Steam sterilization
Specifications
Specifications
Autoclavable
For sterilization
Emulating Indicator Strip
When placed inside trays, packs or pouches, a TST Control Emulating Indicator will confirm that good quality steam has penetrated to the point of placement
100 Pk.
3.5 min.
ISO 11140 part 1, class 6
Autoclave
134°C
Paper
Yes
Steam