missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JULABO™ Tube Fitting Adapters
Tube Fitting Adapters are available in various styles to address laboratory needs.
Brand: JULABO™ 8890105
This item is not returnable.
View return policy
Description
- Adapter fitting styles include:
- M24x1.5 male to 12 mm barbed fittings
- G 1 in. male to G 3/4 in. female
- G 1 1/2 in. male to G 3/4 in. female
- G 1 in. male to G 3/4 in. male
- G 1 1/2 in. male to G 3/4 in. male
Specifications
G 1 in. Male to G 3/4 in. Female | |
Fitting Adapter, Fluid Tubing | |
2 Pk. |