Learn More
Kartell™ Hoses Selector
Kartell™ Hoses Selector provides the nominal outside and inside diameter of hoses, allowing one to determine grip for connectors.
Brand: Kartell™ 0175912
This item is not returnable.
View return policy
Description
- Hoses selector gives nominal O.D. and I.D. of hoses
- Use to determine grip for connectors
- Selector includes following inside diameter in millimeters
- 1, 2, 2.5, 3, 4, 4.5, 5, 6, 7, 8, 9.5, 10, 11, 12, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 25
- Selector includes following outside diameter in millimeters
- 1.5, 2, 2.5, 3, 4, 5.5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, 16, 17, 18, 21
Specifications
ABS | |
Hoses | |
Green |
165 mm | |
90 mm | |
Tube Selector |