Learn More
Fisherbrand™ Tube-Shaped Stirrer Bars
Designed for use with 10mm standard cuvettes. Fisherbrand™ Tube-Shaped Stirrer Bars are isostatically molded in pure FDA-approved PTFE plastic.
Brand:  Fisherbrand™ 001.1609
Product Code. 11818902
  • 16.60 € / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guaranteed inert and leak-free
  • Available with Alnico V or rare earth magnets
  • Individually polished to stay cleaner for longer
Specifications
Specifications
Autoclavable
White
9 x 9 x 6mm
6mm
9 mm
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
Cuvettes
9 mm
5/Pk.