missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Tubed Media for Sartorius Membrane Filtration Units: VLB-S7-S Broth

Tubed media provides time and cost savings

Brand:  Sartorius 14127----------K

266.69 EUR valid until 2024-12-31
Use promo code "21974" to get your promotional price.



Product Code. 10612592

  • 329.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in tubes are for pouring agar plates. The content of one tube is sufficient for two 90mm or three 60mm Petri dishes.

  • 20mL tubes, 50 tubes per box
  • According to international formulas
  • Consistent quality

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
VLB-S7-S Broth
Determination of LactoBacilli and Pediococci and other beer spoiling organisms
20 mL
Germany
20 mL
Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X50 20ML MEDIA VLB-S7-S ST

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers