Learn More
Thermo Scientific™ Tubes and Domed Caps, strips of 8
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Brand: Thermo Scientific™ AB0266
155.48 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights- Compatible with 0.2 mL thermal cycler blocks 8 tubes per strip 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips
- Ultra-Clear cap options available for use in qPCR assays
- PCR
- qPRC
- Real-Time PCR

Specifications
Certified DNA/RNase Free | |
0.2 mL | |
250 Strips | |
Non-sterile | |
Standard | |
96 | |
8-Strip Tube |
Natural | |
0.25 mL | |
Thermal Cyclers | |
Polypropylene | |
Domed Cap | |
250 Strip Tubes and Caps |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.