missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Tubetest COD Reagent Tube with Mercury
Practical and convenient method for testing Chemical Oxygen Demand (COD) using the Palintest™ COD Tubetests system.
89.65 € - 90.65 €
Specifications
For Use With (Equipment) | Photometer or UV-Vis Spectrophotometer |
---|---|
Includes | Chemical Oxygen Demand (COD) |
No. of Tests | 25 |
Type | COD Reagent Tube |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
11928937
|
Palintest™
PL465 |
0 to 2000mg/L |
89.65 €
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11908937
|
Palintest™
PL462 |
0 to 400mg/L |
90.65 €
0.72 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The Palintest™ COD Tubetests system offers a method which is both practical and convenient
- The reagents are supplied premixed and with a minimum of potentially hazardous chemicals
- The test is simply carried out by adding the sample to the tube, closing the cap and then digesting for two hours at 150°C
- The tubes are then cooled and the test colour measured using a Palintest™ photometer
Specifications
Photometer or UV-Vis Spectrophotometer | |
Chemical Oxygen Demand (COD) | |
25 | |
COD Reagent Tube |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COD 2000/2M Solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H301 + H311-Toxic if swallowed or in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P314-Get medical advice/attention if you feel unwell.
- P321-Specific treatment (see on this label).
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P363-Wash contaminated clothing before reuse.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, mercuric sulphate, silver sulphate, potassium dichromate