missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Tubetest Mercury Free COD Reagent Tube

This mercury-free COD reagent tube is ideal for measuring low chloride samples such as Chemical Oxygen Demand (COD).

Brand:  Palintest™ PL456


 View more versions of this product

Product Code. 11923727

  • 179.00 € / Pack of 24

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Mercury-free COD reagent tube for measuring low chloride samples such as Chemical Oxygen Demand (COD)
  • For use with Photometer or UV-Vis Spectrophotometer equipment
  • Type: COD Reagent Tube 
  • Range: 0 to 20000mg/L
Specifications

Specifications

Low Chloride Samples Such as Chemical Oxygen Demand (COD)
Chemical Oxygen Demand (COD)
0 to 20000mg/L
Photometer or UV-Vis Spectrophotometer
25
COD Reagent Tube
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD 2000 Solution

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric aicd, silver sulphate, potassium dichromate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers