Learn More
Tubing Holder
Polypropylene clip captures and holds flexible tubing firmly in place in beakers or other open containers.
Brand:  Bel-Art - SP Scienceware F18235-0000
Product Code. 10527551
  • 35.01 € / Pack of 6
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fits snugly over edge of beaker
  • Long arm measures 2 in. (51mm) with hole to accept tubing up to 0.375 in. (9.5mm) O.D.
  • Autoclavable
Specifications
Specifications
Tubing Holder
Autoclavable
Polypropylene