missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Tubocurarine chloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Competitive, non-selective nicotinic acetylcholine receptor antagonist; causes skeletal muscle relaxation. Also a 5-HT3 and GABAA receptor antagonist.
Specifications
Specifications
CAS | 57-94-3 |
Molecular Formula | C37H43Cl2N2O6+ |
Quantity | 50 mg |
Synonym | tubocurarine chloride |
InChI Key | GXFZCDMWGMFGFL-KKXMJGKMSA-O |
SMILES | CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C6C(CC7=CC(=C(C=C7)O)O3)[N+](CCC6=CC(=C5O)OC)(C)C)OC.Cl.Cl |
Molecular Weight (g/mol) | 682.659 |
PubChem CID | 129894468 |
Formula Weight | 681.65 |
Percent Purity | 0.98 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'