missing translation for 'onlineSavingsMsg'
Learn More

Tungsten boride, 99% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 12007-09-9
Molecular Formula BW
Molecular Weight (g/mol) 194.65
MDL Number MFCD00049708
InChI Key JEEHQNXCPARQJS-UHFFFAOYSA-N
Synonym tungsten boride, tungsten monoboride, tungstenioborane
PubChem CID 82793
IUPAC Name boranylidynetungsten
SMILES B#[W]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327407
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012559.14
25 g N/A
N/A
11337407
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012559.22
100 g N/A
N/A
Description

Description

it is used as an abrasives used in thin, thick Film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12007-09-9
194.65
JEEHQNXCPARQJS-UHFFFAOYSA-N
82793
B#[W]
BW
MFCD00049708
tungsten boride, tungsten monoboride, tungstenioborane
boranylidynetungsten

Specifications

12007-09-9
BW
tungsten boride, tungsten monoboride, tungstenioborane
JEEHQNXCPARQJS-UHFFFAOYSA-N
boranylidynetungsten
82793
99%
(metals basis)
Powder
MFCD00049708
It is insoluble in water.
B#[W]
194.65
194.66
Odorless
Tungsten boride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-498-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only