Learn More
Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼2800°C |
Color | Black |
Physical Form | Powder |
Quantity | 250 g |
Linear Formula | W2C |
UN Number | UN3178 |
Solubility Information | Soluble in nitric acid and hydrofluoric acid. Insoluble in water. |
Formula Weight | 379.71 |
Percent Purity | 99.5% |
Density | 17.15 g/mL |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.