missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99% (metals basis)

Product Code. p-4782215
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11328488

Brand: Thermo Scientific Alfa Aesar 040502.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-12-1 | CW | 195.85 g/mol

Sintered tungsten carbide cutting tools are very abrasion resistant. Tungsten carbide is often used in armor-piercing ammunition. Tungsten carbide is also an effective neutron reflector. Hard carbides, especially tungsten carbide, are used by athletes for balance. Tungsten carbide is also used for making surgical instruments

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-12-1
Molecular Formula CW
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011464
InChI Key UONOETXJSWQNOL-UHFFFAOYSA-N
Synonym tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
IUPAC Name tungsteniomethanide
SMILES [C-]#[W+]

Specifications

Melting Point 2870°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%, metal basis
Solubility Information Insoluble in water. Soluble in nitric acid and hydrochloric acid.
Formula Weight 195.86
Density 15.63
Chemical Name or Material Tungsten Carbide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.