Learn More
Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
83.70 € - 1020.00 €
Chemical Identifiers
CAS | 12036-22-5 |
---|---|
Molecular Formula | O2W |
Molecular Weight (g/mol) | 215.84 |
MDL Number | MFCD00049709 |
InChI Key | DZKDPOPGYFUOGI-UHFFFAOYSA-N |
Synonym | tungsten dioxide, tungsten iv oxide, tungsten oxide wo2, acmc-1bqvs, tungsten iv oxide-w, tungsten iv oxide trace metals basis 5g, tungsten iv oxide,-100 mesh trace metals basis |
PubChem CID | 82850 |
IUPAC Name | dioxotungsten |
SMILES | O=[W]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11313938
|
Thermo Scientific Alfa Aesar
040367.06 |
5 g | N/A | N/A | N/A | |||||
11323938
|
Thermo Scientific Alfa Aesar
040367.14 |
25 g | N/A | N/A | N/A | |||||
11333938
|
Thermo Scientific Alfa Aesar
040367.22 |
100 g | N/A | N/A | N/A | |||||
Description
Tungsten(IV) oxide is used in the production of tungsten and tungstates, in alloys, in fire-proofing fabrics and as a ceramic pigment. It is used to study their chromism, photocatalysis and sensing capabilities. It is associated with rhenium trioxide particles and used for detection of protonatable, radical and precharged species under laser desorption ionization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-22-5 | |
215.84 | |
DZKDPOPGYFUOGI-UHFFFAOYSA-N | |
82850 | |
O=[W]=O |
O2W | |
MFCD00049709 | |
tungsten dioxide, tungsten iv oxide, tungsten oxide wo2, acmc-1bqvs, tungsten iv oxide-w, tungsten iv oxide trace metals basis 5g, tungsten iv oxide,-100 mesh trace metals basis | |
dioxotungsten |
Specifications
1500°C to 1600°C (in N2) | |
Powder | |
O2W | |
tungsten dioxide, tungsten iv oxide, tungsten oxide wo2, acmc-1bqvs, tungsten iv oxide-w, tungsten iv oxide trace metals basis 5g, tungsten iv oxide,-100 mesh trace metals basis | |
DZKDPOPGYFUOGI-UHFFFAOYSA-N | |
dioxotungsten | |
82850 | |
10.8 |
12036-22-5 | |
99.9% (metals basis) | |
MFCD00049709 | |
Soluble in acid and potassium hydroxide. Insoluble in water. | |
O=[W]=O | |
215.84 | |
215.85 | |
Tungsten(IV) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 234-842-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.