missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten(IV) selenide, 99.8% (metals basis)
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
Tungsten(IV) selenide is used as an important compound in electrochemical solar cells. It acts as a stable semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.8%, metal basis |
| Solubility Information | Insoluble in water. |
| Formula Weight | 341.77 |
| Density | 9.32 |
| Chemical Name or Material | Tungsten(IV) Selenide |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu