missing translation for 'onlineSavingsMsg'
Learn More

Tungsten oxide, 99.99% (metals basis)

Product Code. p-4904564
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 11463870

Brand: Thermo Scientific Alfa Aesar 089949.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12037-58-0 | O2W | 215.84 g/mol

Tungsten oxide is used as a pigment in ceramics and paints. It is used in various types of infrared absorbing coatings and foils. It is involved in the production of electrochromic windows, which is an electrically switchable glass that change light transmission properties with an applied voltage. It is a unique material utilized for various applications due their chromism, photocatalysis, and sensing capabilities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-58-0
Molecular Formula O2W
Molecular Weight (g/mol) 215.84
MDL Number MFCD00049709
InChI Key DZKDPOPGYFUOGI-UHFFFAOYSA-N
IUPAC Name dioxotungsten
SMILES O=[W]=O

Specifications

Melting Point 1500°C to 1600°C
Color Blue to Brown
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% metal basis
Solubility Information Soluble in hot alkalis. Insoluble in water and acids.
Formula Weight 230.25
Density 7.16
Chemical Name or Material Tungsten oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.