missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten Rhenium wire, 0.25mm (0.01in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
97.69 € - 425.12 €
Specifications
Chemical Name or Material | Tungsten Rhenium wire |
---|---|
Name Note | 0.25mm (0.01 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | W:Re; 97:3 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD02091751 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336777
|
Thermo Scientific Alfa Aesar
010073.G2 |
2 m |
97.69 €
2m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326777
|
Thermo Scientific Alfa Aesar
010073.G6 |
10 m |
425.12 €
10m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten Rhenium wire | |
W:Re; 97:3 wt% | |
MFCD02091751 | |
≈1.9g/2m |
0.25mm (0.01 in.) dia. | |
(metals basis) | |
99.95% | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title