missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(V) dichloride triethoxide, 10% w/v in ethanol, Thermo Scientific™

Product Code. 11343548
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45000-93-9
Molecular Formula C6H18Cl2O3W
Molecular Weight (g/mol) 392.947
MDL Number MFCD00210651
InChI Key AQDCOTYKQNGVLX-UHFFFAOYSA-L
Synonym tungsten v dichloride triethoxide w/v in ethanol
PubChem CID 86208842
IUPAC Name dichlorotungsten;ethanol
SMILES CCO.CCO.CCO.Cl[W]Cl

Specifications

Color Blue
Physical Form Liquid
Quantity 25 mL
Linear Formula WCl2(OC2H5)3
UN Number UN2924
Formula Weight 389.94
Concentration or Composition (by Analyte or Components) 10% w/v in ethanol
Odor Alcohol-like
Sensitivity Moisture sensitive
Chemical Name or Material Tungsten(V) dichloride triethoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.