missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals

52.76 € - 939.68 €

Chemical Identifiers

CAS 13283-01-7
Molecular Formula Cl6W
Molecular Weight (g/mol) 396.54
MDL Number MFCD00011463
InChI Key KPGXUAIFQMJJFB-UHFFFAOYSA-H
Synonym tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
PubChem CID 83301
ChEBI CHEBI:37771
IUPAC Name hexachlorotungsten
SMILES Cl[W](Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11301709
SDS View Documents
Thermo Scientific Alfa Aesar
012337.06
5 g
52.76 €
5g
Please to purchase this item. Need a web account? Register with us today!
11371699
SDS View Documents
Thermo Scientific Alfa Aesar
012337.09
10 g
74.67 €
10g
Please to purchase this item. Need a web account? Register with us today!
11391699
SDS View Documents
Thermo Scientific Alfa Aesar
012337.18
50 g
261.30 €
50g
Please to purchase this item. Need a web account? Register with us today!
11381699
SDS View Documents
Thermo Scientific Alfa Aesar
012337.30
250 g
939.68 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a precursor for many tungsten compounds. It is used in tungsten plating, purification of tungsten, and olefin polymerization. It is also used as petroleum catalyst, and in intelligent glass. It can be used for deoxygenation of epoxides and as a reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13283-01-7
396.54
KPGXUAIFQMJJFB-UHFFFAOYSA-H
83301
hexachlorotungsten
Cl6W
MFCD00011463
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
CHEBI:37771
Cl[W](Cl)(Cl)(Cl)(Cl)Cl

Specifications

346°C
13283-01-7
99%
MFCD00011463
Soluble in carbon disulfide,carbon tetrachloride and phosphorus oxychloride.
Cl[W](Cl)(Cl)(Cl)(Cl)Cl
396.54
CHEBI:37771
Moisture sensitive
Tungsten(VI) Chloride
275°C
Powder
Cl6W
tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
KPGXUAIFQMJJFB-UHFFFAOYSA-H
hexachlorotungsten
83301
396.57
3.52
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c

H302-H314

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 236-293-9

missing translation for 'rtecsNumber' : YO7710000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.