missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) oxide, 99+%, Thermo Scientific Chemicals

77.20 € - 252.00 €

Chemical Identifiers

CAS 1314-35-8
MDL Number MFCD00011466
InChI Key ZNOKGRXACCSDPY-UHFFFAOYSA-N
Synonym tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
PubChem CID 14811
IUPAC Name trioxotungsten
SMILES O=[W](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10042750
View Documents Promotion Details
Thermo Scientific Acros
214211000
100 g Plastic bottle
77.20 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10549823
View Documents Promotion Details
Thermo Scientific Acros
214215000
500 g Plastic bottle
252.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-35-8
ZNOKGRXACCSDPY-UHFFFAOYSA-N
14811
O=[W](=O)=O
MFCD00011466
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
trioxotungsten

Specifications

1314-35-8
Powder
O3W
MFCD00011466
15, 9997
ZNOKGRXACCSDPY-UHFFFAOYSA-N
trioxotungsten
14811
≥99.0%
Green-Yellow to Yellow
99+%
WO3
11,591
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
O=[W](=O)=O
231.837
231.85
Tungsten(VI) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Identifier
  • Tungsten(VI) oxide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only