missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Wire, 0.05mm (0.002 in.) dia., annealed, 99.95% (Metals basis), Thermo Scientific Chemicals

92.16 € - 847.00 €

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369688
View Documents
Thermo Scientific Alfa Aesar
044191.H4
100 m
92.16 €
100m
Please to purchase this item. Need a web account? Register with us today!
11379688
View Documents
Thermo Scientific Alfa Aesar
044191.H9
500 m
293.46 €
500m
Please to purchase this item. Need a web account? Register with us today!
11389688
View Documents
Thermo Scientific Alfa Aesar
044191.I2
2500 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]

Specifications

Silver to Gray
Silver Gray wire
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Tungsten, Annealed
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-143-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.