missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, 0.1mm (0.004in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11365888
Click to view available options
Quantity:
250 m
4 x 250 m
50 m

Form: Approx. 0.15g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifica

Color Silver to Gray
Appearance Silver Gray wire
Density 19.3g/cm3
Assay Percent Range 99.95% (Metals basis)
Quantity 50 m
Form Wire
Chemical Name or Material Tungsten

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'