missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041039.G6
543.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Tungsten wire, gold plated | |
(metals basis) | |
10 m | |
Au 99.99% | |
Wire |
0.05mm (0.002 in.) dia. | |
MFCD00011461 | |
99.95% | |
≈0.02g/m (W only), 3 to 5 wt% Au plating |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction