Learn More
Tungsten wire, gold plated, 0.1mm (0.004 in.) dia., W 99.95% (metals basis), Au 99.99%, Thermo Scientific Chemicals
131.66 € - 610.00 €
Specifications
Chemical Name or Material | Tungsten wire, gold plated |
---|---|
Name Note | 0.1mm (0.004 in.) dia. |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00011461 |
Percent Purity | 99.95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15493935
|
Thermo Scientific Alfa Aesar
045086.G1 |
1 m |
131.66 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403945
|
Thermo Scientific Alfa Aesar
045086.G5 |
5 m |
399.99 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15413945
|
Thermo Scientific Alfa Aesar
045086.G6 |
10 m | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten wire, gold plated | |
(metals basis) | |
99.95% | |
≈0.15g/m (W only), 3 to 5 wt% Au plating |
0.1mm (0.004 in.) dia. | |
MFCD00011461 | |
Au 99.99%, W 99.95% | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.