missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, platinum coated, 0.25mm (0.01in) dia, Thermo Scientific Chemicals

191.96 € - 794.96 €

Specifications

Chemical Name or Material Tungsten wire, platinum coated
Name Note 0.25mm (0.01 in.) dia.
MDL Number MFCD00011461
Particle Size thickness 5 Micron
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336888
View Documents
Thermo Scientific Alfa Aesar
1 m
191.96 €
1m
Please to purchase this item. Need a web account? Register with us today!
11326888
View Documents
Thermo Scientific Alfa Aesar
5 m
794.96 €
5m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tungsten wire, platinum coated
0.25mm (0.01 in.) dia.
MFCD00011461
thickness 5 Micron
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only