missing translation for 'onlineSavingsMsg'
Learn More

Tungstic acid, 99+%, Thermo Scientific Chemicals

23.80 € - 240.20 €

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.86
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
IUPAC Name dioxotungstendiol
SMILES O[W](O)(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10318300
View Documents
Thermo Scientific Acros
207610050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10134740
missing translation for 'viewDocuments'
Thermo Scientific Acros
207611000
100 g Plastic bottle
66.53 €
100g
Please to purchase this item. Need a web account? Register with us today!
10107423
missing translation for 'viewDocuments'
Thermo Scientific Acros
207615000
500 g Plastic bottle
240.20 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7783-03-1
249.86
CMPGARWFYBADJI-UHFFFAOYSA-L
O[W](O)(=O)=O
H2O4W
MFCD00011336
dioxotungstendiol

Specifications

7783-03-1
Powder
H2O4W
MFCD00011336
Solubility in water: insoluble. Other solubilities: soluble in hf and alkalies
O[W](O)(=O)=O
249.86
≥99%
5.5000g/mL
Green-Yellow to Yellow
99% min
H2WO4
15, 9998
CMPGARWFYBADJI-UHFFFAOYSA-L
dioxotungstendiol
249.86
5.5
Tungstic acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-975-2

Product Identifier
  • Tungstic acid

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.