missing translation for 'onlineSavingsMsg'
Learn More

Tungstic(VI) acid, Thermo Scientific Chemicals

43.50 € - 329.00 €

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.85
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
Synonym tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
PubChem CID 1152
ChEBI CHEBI:36272
IUPAC Name dihydroxy(dioxo)tungsten
SMILES O[W](=O)(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364929
View Documents Promotion Details
Thermo Scientific Alfa Aesar
082118.14
25 g
43.50 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11354929
View Documents Promotion Details
Thermo Scientific Alfa Aesar
082118.22
100 g
72.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11374929
View Documents Promotion Details
Thermo Scientific Alfa Aesar
082118.36
500 g
329.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applied as flavoring agent and fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-03-1
249.85
CMPGARWFYBADJI-UHFFFAOYSA-L
1152
dihydroxy(dioxo)tungsten
H2O4W
MFCD00011336
tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
CHEBI:36272
O[W](=O)(=O)O

Specifications

1473°C
7783-03-1
H2O4W
tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
CMPGARWFYBADJI-UHFFFAOYSA-L
dihydroxy(dioxo)tungsten
1152
249.86
5.5
100°C (decomposition)
Powder
MFCD00011336
Insoluble in water and acid. Soluble in alkaline solution.
O[W](=O)(=O)O
249.85
CHEBI:36272
2.24
Tungstic(VI) acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-975-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only