missing translation for 'onlineSavingsMsg'
Learn More

Tungstosilicic acid hydrate, Reagent Grade, Thermo Scientific Chemicals

50.45 € - 132.66 €

Chemical Identifiers

CAS 12027-43-9
Molecular Formula H4O40SiW12
Molecular Weight (g/mol) 2878.16
MDL Number MFCD00036271
InChI Key HIKIIEGSHYIJCC-UHFFFAOYSA-K
Synonym tungstosilicic acid, tungstosilicic acid hydrate, p.a., tungstosilicic acid hydrate, purum p.a., tungstosilicic acid hydrate, reagent grade, tungstosilicic acid hydrate trace metals basis, tungstate 4-, m12-orthosilicato 4--ko:ko:ko:ko':ko':ko':ko:ko:ko:ko':ko':ko' tetracosa-m-oxododecaoxododeca-, hydrogen 1:4
PubChem CID 71307034
SMILES O[Si](O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)(O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324607
SDS View Documents
Thermo Scientific Alfa Aesar
039651.09
10 g
50.45 €
10g
Please to purchase this item. Need a web account? Register with us today!
11334607
SDS View Documents
Thermo Scientific Alfa Aesar
039651.18
50 g
132.66 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalyst for organic synthesis, minerals separation, reagent for alkaloids.Tungstosilicic acid hydrate is used to prepare ethylacetate and acetic acid from ethylene. It is used in the preparation of silicotungstate butylmethylimidazolium bromide and charge transfer compounds with Keggin anions. It is also used in catalyst preparation, which is useful for catalytic liquid-phase oxidation of xylene. Further, it acts as a mediator in production of hydrogen through electrolysis of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12027-43-9
2878.16
HIKIIEGSHYIJCC-UHFFFAOYSA-K
71307034
H4O40SiW12
MFCD00036271
tungstosilicic acid, tungstosilicic acid hydrate, p.a., tungstosilicic acid hydrate, purum p.a., tungstosilicic acid hydrate, reagent grade, tungstosilicic acid hydrate trace metals basis, tungstate 4-, m12-orthosilicato 4--ko:ko:ko:ko':ko':ko':ko:ko:ko:ko':ko':ko' tetracosa-m-oxododecaoxododeca-, hydrogen 1:4
O[Si](O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)(O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O

Specifications

12027-43-9
H4O40SiW12
tungstosilicic acid, tungstosilicic acid hydrate, p.a., tungstosilicic acid hydrate, purum p.a., tungstosilicic acid hydrate, reagent grade, tungstosilicic acid hydrate trace metals basis, tungstate 4-, m12-orthosilicato 4--ko:ko:ko:ko':ko':ko':ko:ko:ko:ko':ko':ko' tetracosa-m-oxododecaoxododeca-, hydrogen 1:4
HIKIIEGSHYIJCC-UHFFFAOYSA-K
9-[(7-hydroxy-1,1,3,3,5,5,7,7-octaoxotetratungstoxan-1-yl)oxy]-1,1,3,3,5,5,7,7,11,11,13,13,15,15,17,17-hexadecaoxo-2,4,6,8,10,12,14,16-octaoxa-9-sila-1,3,5,7,11,13,15,17-octatungstaheptadecane-1,9,17-triol
71307034
Reagent
Crystalline
MFCD00036271
Soluble in water.
O[Si](O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)(O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O
2878.16
2878.28 (anhydrous)
Tungstosilicic acid hydrate, Reagent Grade
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.